Order Entry
Slovakia
ContactUsLinkComponent
Recombinant Treponema pallidum p41
Recombinant Treponema pallidum p41
Catalog # APOSBITP1858-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Treponema pallidum p41
Catalog # APOSBITP1858-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Treponema p41
  • Protein/peptide name:
    Treponema pallidum p41

Specifications

About this item