Order Entry
Slovakia
ContactUsLinkComponent
 
Recombinant Hepatitis A Virus VP3
Catalog # APOSBITP1818-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis A Virus VP3
Catalog # APOSBITP1818-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HAV VP3
  • Protein/peptide name:
    Hepatitis A Virus VP3

Specifications

About this item