0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Proteins and Peptides
Recombinant Hepatitis C Virus NS4 a+b
Recombinant Hepatitis C Virus NS4 a+b
Recombinant Hepatitis C Virus NS4 a+b
Catalog # APOSBITP1791-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant Hepatitis C Virus NS4 a+b
Catalog # APOSBITP1791-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HCV NS4 a+b
Protein/peptide name:
Hepatitis C Virus NS4 a+b
Specifications
About this item
Product Details
Product Details