Order Entry
Slovakia
ContactUsLinkComponent
Recombinant Herpes Simplex Virus-2 gD
Recombinant Herpes Simplex Virus-2 gD
Catalog # APOSBITP1738-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Herpes Simplex Virus-2 gD
Catalog # APOSBITP1738-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HSV-2 gD
  • Protein/peptide name:
    Herpes Simplex Virus-2 gD

Specifications

About this item