
3,5-Difluoro-4-pyridinamine 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:159783-22-9
:
Formula:C₅H₄F₂N₂
MDL Number:MFCD01862101
:130.1 g/mol
Melting Pt:99…101 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together