Order Entry
Slovakia
ContactUsLinkComponent
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Irritant
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Catalog # APOSPC200013-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Catalog # APOSPC200013-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.61 g/mol
Melting Pt:47…49 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1407516-45-3
Formula:C₉H₇ClF₃NO₂
MDL Number:MFCD22573527