Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitrobenzenediazonium tetrafluoroborate
4-Nitrobenzenediazonium tetrafluoroborate 98%
4-Nitrobenzenediazonium tetrafluoroborate 98%
APOSPC10103-25G
:
Apollo Scientific
:
4-Nitrobenzenediazonium tetrafluoroborate 98%
APOSPC10103-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
236.92 g/mol
:
144…148 °C (dec.)
:
Refrigerator
:
207-261-1
:
3261
:
8,II
:
GHS05
:
GHS07
:
456-27-9
:
C₆H₄BF₄N₃O₂
:
MFCD00012005