
Pyrimidine-5-carboxylic acid 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4595-61-3
:
Formula:C5H4N2O2
MDL Number:MFCD00856162
:124.10 g/mol
Melting Pt:259…261 °C
EINECS:224-994-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together