Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminobenzenesulphonic acid
2-Aminobenzenesulphonic acid 95%
2-Aminobenzenesulphonic acid 95%
2-Aminobenzenesulphonic acid 95%
APOSOR40577-1KG
:
Apollo Scientific
:
2-Aminobenzenesulphonic acid 95%
APOSOR40577-1KG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 kg
Seq:
4024
:
:
173.19 g/mol
:
>300 °C
:
>300 °C
:
Refrigerator
:
2585
:
8,III
:
GHS05
:
88-21-1
:
C₆H₇NO₃S
:
MFCD00007705