Bromonitromethane 90%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:563-70-2
- Formula:BrCH₂NO₂
- MDL Number:MFCD00007401
- Molecular Weight:139.94 g/mol
- Boiling Pt:146…148 °C/750mmHg
- Density:2.007 g/cm³ (25 °C)
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:209-258-0
- UN:3139
- ADR:5.1,II
- :Density: 2.007 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
- CAS Number:563-70-2
- Formula:BrCH₂NO₂
- MDL Number:MFCD00007401
- Molecular Weight:139.94 g/mol
- Boiling Pt:146…148 °C/750mmHg
- Density:2.007 g/cm³ (25 °C)
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:209-258-0
- UN:3139
- ADR:5.1,II
- :Density: 2.007 g/cm³ (20 °C)



