Cyanomethyl ethanethioate 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: S-(Cyanomethyl) thioacetate
CAS Number:59463-56-8
:
Formula:C₄H₅NOS
MDL Number:MFCD00173920
Boiling Pt:226…227 °C
Density:1.176 g/cm³ (25 °C)
Molecular Weight:115.15 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together