
Tetrabutylammonium acetate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10534-59-5
:
Formula:C₁₈H₃₉NO₂
MDL Number:MFCD00043208
:301.51 g/mol
Melting Pt:95…98 °C
EINECS:234-101-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together