
2-Bromo-4-methoxyphenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17332-11-5
:
Formula:C₇H₇BrO₂
MDL Number:MFCD02113712
:203.04 g/mol
Boiling Pt:115 °C
Melting Pt:45 °C
Storage Temperature:Ambient
EINECS:605-671-9
:GHS07
Frequently Bought Together