Zadaní objednávky
Slovakia
ContactUsLinkComponent
Tetrakis(dimethylamido)titanium(IV) 99.99%
Flammable
Corrosive
Toxic
Tetrakis(dimethylamido)titanium(IV) 99.99%
Catalog # APOSIN7556-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Toxic
Tetrakis(dimethylamido)titanium(IV) 99.99%
Catalog # APOSIN7556-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:50 °C/0.5mmHg
Melting Pt:<4 °C
Density:0.95 g/cm³ (25 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:221-904-3
UN:3398
ADR:4.3,I
 :224.17 g/mol
 :GHS02
 :GHS05
 :GHS06
CAS Number:3275-24-9
Formula:C₈H₂₄N₄Ti
MDL Number:MFCD00014861