Order Entry
Slovakia
ContactUsLinkComponent
Palladium(II) chloride 98%
Corrosive
Irritant
Palladium(II) chloride 98%
Catalog # APOSIN2804-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Palladium(II) chloride 98%
Catalog # APOSIN2804-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:678…680 °C
Density:4 g/cm³ (25 °C)
 :177.33 g/mol
EINECS:231-596-2
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-10-1
Formula:PdCl₂
MDL Number:MFCD00003558