Antimony(III) chloride 99.99%
Catalog # APOSIN1111-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:228.12 g/mol
Boiling Pt:283 °C
Melting Pt:73.4 °C
Density:3.14 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:10025-91-9
Formula:SbCl₃
MDL Number:MFCD00011212