Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate
Ammonium sulphate 99.999%
Ammonium sulphate 99.999%
Ammonium sulphate 99.999%
APOSIN1084-250G
:
Apollo Scientific
:
Ammonium sulphate 99.999%
APOSIN1084-250G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 g
Seq:
3545
:
:
132.14 g/mol
:
235 °C (dec.)
:
1.769 g/cm³ (25 °C)
:
Ambient
:
7783-20-2
:
(NH₄)₂SO₄
:
MFCD00003391