
Dimethyl sulphoxide, Cell Culture Grade
Catalog # APOSBID1200-4X250M
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:78.14 g/mol
Boiling Pt:189 °C
Melting Pt:18 °C
Density:1.1 g/cm³ (25 °C)
Flash Pt:95 °C
Storage Temperature:Ambient
EINECS:200-604-3
:GHS07
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089