Human recombinant CD7 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Llama IgG2b Fc Tag
- Storage conditions:–20 °C, avoid light
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P09564-1
- Protein synonyms:LEU-9|TP41|CD7|Tp40|GP40
- Protein/peptide name:CD7
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:44,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together