1H,1H,2H,2H-Perfluoro(10,10-dimethyl-1-iodo-9-oxa)decane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25080-25-5
:
Formula:C₁₁H₄F₁₉IO
MDL Number:MFCD09763609
:640.023 g/mol
Storage Temperature:Ambient
Frequently Bought Together