

Perfluoro-3,5,7,8-tetrachlorooctanoic acid 95%
Catalog # APOSPC6855-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:479.887 g/mol
Boiling Pt:154…156.5 °C (10 mm Hg)
Density:1.899 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
:GHS07
CAS Number:2923-68-4
Formula:C₈HCl₄F₁₁O₂
MDL Number:MFCD00046513