Order Entry
Slovakia
ContactUsLinkComponent
1-(4-Fluorophenyl)-2-methylpropan-2-ol
1-(4-Fluorophenyl)-2-methylpropan-2-ol
Catalog # APOSPC5535-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorophenyl)-2-methylpropan-2-ol
Catalog # APOSPC5535-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1,1-Dimethyl-2-(4-fluorophenyl)ethan-1-ol
 :
 :168.208 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2928-17-8
Formula:C₁₀H₁₃FO
MDL Number:MFCD00082699
Boiling Pt:105 °C ( 15 mm Hg)
Density:1.1 g/cm³ (25 °C)
Flash Pt:>110 °C