
3-Bromo-2-(trifluoromethyl)aniline
Catalog # APOSPC53158-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.021 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:244246-71-7
Formula:C₇H₅BrF₃N
MDL Number:MFCD13194482