Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,3-difluorobenzylamine hydrochloride
4-Bromo-2,3-difluorobenzylamine hydrochloride 97%
4-Bromo-2,3-difluorobenzylamine hydrochloride 97%
APOSPC500666-250MG
:
Apollo Scientific
:
4-Bromo-2,3-difluorobenzylamine hydrochloride 97%
APOSPC500666-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
258.491 g/mol
:
GHS07
:
Ambient
:
1980086-47-2
:
C₇H₆BrF₂N·ClH
:
MFCD12546123