Order Entry
Slovakia
ContactUsLinkComponent
Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate 95%
Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate 95%
Catalog # APOSOR918390-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate 95%
Catalog # APOSOR918390-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.194 g/mol
Storage Temperature:Ambient
CAS Number:38410-80-9
Formula:C₈H₁₄O₄
MDL Number:MFCD00077933