Order Entry
Slovakia
ContactUsLinkComponent
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Catalog # APOSOR60138-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Catalog # APOSOR60138-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.22 g/mol
Storage Temperature:Ambient
CAS Number:56079-16-4
Formula:C₉H₁₄N₂O₂
MDL Number:MFCD00159644