Order Entry
Slovakia
ContactUsLinkComponent
2-Bromo-7-methoxynaphthalene
2-Bromo-7-methoxynaphthalene
Catalog # APOSOR470815-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-7-methoxynaphthalene
Catalog # APOSOR470815-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.093 g/mol
Storage Temperature:Ambient
CAS Number:200875-36-1
Formula:C₁₁H₉BrO
MDL Number:MFCD17012538