Order Entry
Slovakia
ContactUsLinkComponent
3-(2-Piperidyl)propyl Acetate Acetate
3-(2-Piperidyl)propyl Acetate Acetate
Catalog # APOSOR470597-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Piperidyl)propyl Acetate Acetate
Catalog # APOSOR470597-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :245.315 g/mol
Storage Temperature:Ambient
CAS Number:1427475-23-7
Formula:C₁₀H₁₉NO₂·CH₃CO₂H
MDL Number:MFCD23381117