Order Entry
Slovakia
ContactUsLinkComponent
2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide, tech
2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide, tech
Catalog # APOSOR33155-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide, tech
Catalog # APOSOR33155-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :252.7 g/mol
Storage Temperature:Ambient
CAS Number:866131-79-5
Formula:C₁₁H₁₃ClN₄O
MDL Number:MFCD03305741