Order Entry
Slovakia
ContactUsLinkComponent
4-Butyl-2,7,7-trimethyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
4-Butyl-2,7,7-trimethyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
Catalog # APOSOR32133-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Butyl-2,7,7-trimethyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
Catalog # APOSOR32133-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.385 g/mol
Storage Temperature:Ambient
CAS Number:860611-69-4
Formula:C₁₇H₂₄N₂O
MDL Number:MFCD04124163