Order Entry
Slovakia
ContactUsLinkComponent
2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
Catalog # APOSOR32132-1G
Supplier:  Apollo Scientific
CAS Number:  
2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
Catalog # APOSOR32132-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :292.375 g/mol
Storage Temperature:Ambient
CAS Number:19732-57-1
Formula:C₁₉H₂₀N₂O
MDL Number:MFCD00524746