N-Boc-PEG6-alcohol
Bottle, 500 mg
Catalog # APOSBIPG1663-500MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 17-(Boc-amino)-3,6,9,12,15-pentaoxa-1-heptadecanol
- Formula:C₁₇H₃₅NO₈
- MDL Number:MFCD28385472
- Molecular Weight:381.462 g/mol
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



