Overalls with hood, Tyvek® 500 HP , model TY198S
DuPont™ Tyvek® 500 HP harness protection with ergonomic-protective design with 2-piece hood.
- Stitched internal seams
- Hook and loop closure when connecting to the carabiner
- Pass-thru system to protect the rope lanyard
- Snap closure for full tightness when pass-thru is not used
- Sealable chin flap & zipper flaps, Tyvek® zipper and flap
- Elasticated wrists, ankles and face, elasticated waist (glued-in), thumb loop
The Tyvek® 500 HP offers the wearer full protection by housing the rope lanyard within a sealed compar™ent giving access to the harness worn underneath, maintaining a failsafe barrier between the user and equipment and any chemical threat.
Extremely lightweight offering high level of comfort and mobility. Free of silicone, oil, grease, contaminates, foreign materials, and surface irregularities (suitable for paint applications).
: Certified according to regulation (EU) 2016/425, category III, Type 5-B and 6-B, EN 14126, EN 1073-2, antistatic treatment (EN 1149-5) - on both sides.
- Catalog No:
- DUPOTY198SWHHP/3XL
- DUPOTY198SWHHP/L
- DUPOTY198SWHHP/XL
- DUPOTY198SWHHP/M
- DUPOTY198SWHHP/S
- DUPOTY198SWHHP/2XL
- Model:
- TY 198S
- TY 198S
- TY 198S
- TY 198S
- TY 198S
- TY 198S
- Anti-static:
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical, Industrial
Frequently Bought Together