Hydra square bottle, GL 45, with 4 side necks, DURAN®
Catalog # 215-4548
Supplier: DWK LIFE SCIENCES
Specifications
- Cap Size:GL 45
- Capacity:1000 ml
- Colour:Clear
- W×D×H:94×94×217,5 mm
- Cap colour:Blue
- Neck I-Ø:30 mm
- Pk:1
Specifications
About this item
HPLC 1000 ml purging bottle with five GL 45 inlets.
- Convenient – purging bottle accepts up to five HPLC GL 45 caps
- Safer – capped bottles reduce the risk of volatile organic compound (VOC) escape and spillage
- Compatible - GL 45 threads fit standard HPLC connection caps
- Manufactured from the material of choice for HPLC solvent bottles: Borosilicate 3.3 glass offering a very low inorganic ion extractables profile
- 1 L square space saving shape
- Durable white enamel volume graduations (100 to 800 ml, in 100 ml intervals)
DURAN® bottles are ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents. Increasing the number of GL 45 inlets on the bottles adds convenience: The five GL 45 necks on this purging bottle increases safety and reduces the risk of cross contamination and air intake.
Applications: HPLC purge bottle, reagent supply to liquid handling systems and analysers.
Certifications: Clear DURAN® is a borosilicate 3.3 glass as specified by ISO 3585, and is classified as a 'Type I, Class A low expansion borosilicate glass' as defined by ASTM-E438-1992. Retrace Code, and certificate for manufacturing lot traceability.
Glass Type I according to the requirements of the current United States Pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European Pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese Pharmacopoeia JP <7.01> Test for Glass Containers for Injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, standard deviation of the batch, and the date of issue.
Glass Type I according to the requirements of the current United States Pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European Pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese Pharmacopoeia JP <7.01> Test for Glass Containers for Injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, standard deviation of the batch, and the date of issue.