High-visibility polo shirts, long sleeve, flame resistant, FR77
Supplier: Portwest
Certificates
About this item
The lightweight, modacrylic fabric together with the reflective strips provide the wearer with advanced visibility and protection in hazardous situations.
- Inherent flame resistant qualities will not diminish with washing
- Protection against radiant, convective and contact heat
- Premium sew on flame resistant reflective tape
- Flame resistant ribbed cuff for added safety
- 50+ UPF rated fabric to block 98% of UV rays
- High cotton content for superior comfort
- Concealed button closure
Shell fabric: Modaflame knit 210 g.
Certifications: AS/NZS 1906.4:2010; EN 1149 -5; EN ISO 11612 (A1, B1, C1, F1); IEC 61482-2 IEC 61482-1-2 Class 1; EN ISO 20471 Class 3; ASTM F1959/F1959M-12; CE-CAT III
Specifications
- Catalog No:
- PRTWFR77YERM
- PRTWFR77YERS
- PRTWFR77YERXL
- PRTWFR77YERL
- PRTWFR77YERXXXL
- PRTWFR77YERXXL
- PRTWFR77YER4XL
- Model:
- FR77
- FR77
- FR77
- FR77
- FR77
- FR77
- FR77
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Modacrylic/cotton/anti-static fibres
- Weight:
- 520 g
- 500 g
- 560 g
- 540 g
- 640 g
- 600 g
- 680 g
Specifications
Frequently Bought Together