Order Entry
Slovakia
ContactUsLinkComponent
Chemical resistant gloves, A643 Amber Cut
 :  Portwest
undefined
Chemical resistant gloves, A643 Amber Cut
 :  Portwest
  • Catalog No:
  • PRTWA643AMRM
  • PRTWA643AMRL
  • PRTWA643AMRXL
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyester/elastane
  • Polyester/elastane
  • Polyester/elastane
  • Finish:
  • Palm dipped
  • Palm dipped
  • Palm dipped
  • Gauge:
  • 13
  • 13
  • 13
  • EN norm:
  • EN 388:2016 +A1:2018 (4X42B)
  • EN 388:2016 +A1:2018 (4X42B)
  • EN 388:2016 +A1:2018 (4X42B)
  • Food contact approved:
  • +
  • +
  • +
Frequently Bought Together