About this item
SecureFit™ 400X safety glasses feature 3M™ Pressure Diffusion Temple Technology, wide flexible temple arms, and a combined soft nose bridge and brow guard. These lightweight safety glasses feature a clear, anti-scratch and superior anti-fog, rimless polycarbonate lenses.
- Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
- Wrap-around design improves the security of fit and prevents slipping
- Padded temple touch points for increased comfort over the ears
- Soft nose bridge for a more comfortable fit
- Contoured brow guard provides additional coverage from flying particles
- Rugged anti-scratch coating meets the K marking requirements of EN 166
- Clear lens for excellent protection against ultraviolet (UV) radiation
Lightweight eye protection designed for comfort and performance, these safety glasses weigh just 22 g and have a contemporary, stylish look. The 3M™ Pressure Diffusion Temple Technology ensures a comfortable fit, with wide, flexible temple arms over the ears that provide solid side protection and padding for increased comfort. The soft nose bridge and contoured brow guard rest comfortably against the forehead to help provide additional protection from flying particles, and the wraparound design gives the user a wide field of vision as well as reducing slippage from head movements.
Scotchgard™ anti-fog coating ensures clear visibility, resisting fogging longer than traditional anti-fog coatings even after washing multiple times, including using alcohol wipes or disinfecting by soaking in diluted bleach. Superior anti-fog and anti-scratch properties provide increased durability and visibility. These glasses also combine well with earmuffs.