Order Entry
Slovakia
Orders LinkContactUsLinkComponent
KLEEN blade management system
Supplier:   SWANN MORTON
KLEEN blade management system
Supplier:  SWANN MORTON
Certificates
KLEEN blade management system
KLEEN blade management system
11 Options Available Below
Product Details & Documents
The KLEEN Blade Management System allows for effortless safe attachment and removal of surgical blades from a standard handle. The system which incorporates a quality Swann-Morton surgical blade is designed to operate with most No.3 and No.4 standard surgical handles already in circulation within the hospital system thus minimising disruption and cost during the conversion process.
  • Sterile and non sterile blades
  • Protective sliding sheath
The sheath covers the blade completely thus assisting with accident free handling. It fully exposes the surgical blade when in use with an intuitive lock which prevents any forward movement of the sheath during the procedure. Easy to detach the BMS encapsulates the contaminated sharp which is still visible for the final count and to help ensure safe and responsible disposal.
Product Family Options
Sterile
Product Information
  • Material
    Availability
    Price
  • SWAN5701
    Kleen BMS No. 10
    Kleen BMS No. 10
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 321,82 €
    321,82 €
    1 * 50 items
       321,82 €
  • SWAN5703
    Kleen BMS No. 11
    Kleen BMS No. 11
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 321,82 €
    321,82 €
    1 * 50 items
       321,82 €
  • SWAN5791
    Kleen BMS No. 11P
    Kleen BMS No. 11P
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 321,82 €
    321,82 €
    1 * 50 items
       321,82 €
  • SWAN5705
    Kleen BMS No. 15
    Kleen BMS No. 15
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 321,82 €
    321,82 €
    1 * 50 items
       321,82 €
  • SWAN5792
    Kleen BMS No. 15T
    Kleen BMS No. 15T
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 321,82 €
    321,82 €
    1 * 50 items
       321,82 €
  • SWAN5706
    Kleen BMS No. 20
    Kleen BMS No. 20
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 340,32 €
    340,32 €
    1 * 50 items
       340,32 €
  • SWAN5707
    Kleen BMS No. 21
    Kleen BMS No. 21
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 340,32 €
    340,32 €
    1 * 50 items
       340,32 €
  • SWAN5708
    Kleen BMS No. 22
    Kleen BMS No. 22
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 340,32 €
    340,32 €
    1 * 50 items
       340,32 €
  • SWAN5710
    Kleen BMS No. 23
    Kleen BMS No. 23
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 340,32 €
    340,32 €
    1 * 50 items
       340,32 €
  • SWAN5711
    Kleen BMS No. 24
    Kleen BMS No. 24
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 340,32 €
    340,32 €
    1 * 50 items
       340,32 €
Non sterile
Product Information
  • Material
    Availability
    Price
  • SWAN5606
    Kleen BMS No. 20
    Kleen BMS No. 20
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 728,32 €
    2 728,32 €
    1 * 500 items
       2 728,32 €