
Iron(III) sulphate hydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0123.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (Fe2(SO4)3):min. 73.0 %
- Chloride (Cl):max. 0.002 %
- Copper (Cu):max. 0.005 %
- Zinc (Zn):max. 0.005 %
- Ferrous Iron (as Fe):max. 0.02 %
- Insoluble Matter:max. 0.02 %
- Nitrate (NO3):max. 0.01 %
- Substances not Precipitated by NH4OH (as SO4):max. 0.1 %
Specification Test Results
About this item
:
:399.88 g/mol (anhy)
Merck Index:13,04065
Density:3.1 g/cm³ (20 °C)
EINECS:233-072-9
:GHS07
Storage Temperature:Ambient
CAS Number:15244-10-7
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00149714