Supelco® Organic Reference Standard, (−)-Borneol
15598-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Standards:Analytical standard
- CAS No.:464-45-9
- Description:(−)-Borneol
- Seq:01
- Storage conditions:2...30 °C
- Hazard class description:H228
- Shipping conditions:STD
- MDL No.:MFCD00003759
- M-Clarity quality level:MQ100
- Synonym(s):endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
- Molecular weight:154,25
- Flash point:66 °C
- GHS signal: GHS02, GHS07
: P210 - P240 - P241 - P280 - P370 + P378