Supelco® PESTANAL® Organic Reference Standard, α-Cypermethrin
45806-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:67375-30-8
- Description:α-Cypermethrin
- Storage conditions:2...30 °C
- Hazard class description:H301 - H335 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055328
- M-Clarity quality level:MQ100
- Synonym(s):α-metherin; Alphamethrin
- Molecular weight:416,30
- GHS signal: GHS06, GHS08, GHS09
: P260 - P264 - P271 - P273 - P301 + P310 - P314