Order Entry
Slovakia
ContactUsLinkComponent
Tetrahydrofuran-[D₈], Sigma-Aldrich®
Tetrahydrofuran-[D₈], Sigma-Aldrich®
Catalog # SIAL269891-10G
Supplier:  Merck
CAS Number:  
Tetrahydrofuran-[D₈], Sigma-Aldrich®
Catalog # SIAL269891-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :80.04 g/mol
Boiling Pt:65 °C (1013 hPa)
Melting Pt:–108 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:–17 °C (closed cup)
Storage Temperature:Ambient
EINECS:216-898-4
UN:2056
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
CAS Number:1693-74-9
Formula:C₄D₈O
MDL Number:MFCD00044238