Order Entry
Slovakia
ContactUsLinkComponent
Phenol 99.0-100.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Corrosive
Health
Toxic
Phenol 99.0-100.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier:  HONEYWELL
Certificates
Corrosive
Health
Toxic
Phenol 99.0-100.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier:  HONEYWELL

Specification Test Results

  • Assay (GC):
    99.5 - 100.5 %
  • Assay:
    99.0 - 100.5 %
  • Congealing - Freezing Point:
    min. 40.5 °C
  • Boiling range:
    180 - 181 °C
  • Non-volatile matter:
    max. 0.01 %
  • Water (Karl Fischer):
    max. 0.2 %
  • Iron (Fe):
    max. 0.0001 %
  • Heavy metals (as Pb):
    max. 0.001 %
  • Chloride (Cl):
    max. 0.0005 %
  • o-cresol (GC):
    max. 0.05 %
  • m-Cresol (GC):
    max. 0.05 %
  • p-Cresol (GC):
    max. 0.05 %
  • Sensitivity as microanalytical reagent:
    complying
  • Acidity:
    complying
  • Clearness of the solution:
    complying
  • Appearance of the solution:
    complying

Specification Test Results

Frequently Bought Together