Calcium assay kit
Calcium Assay Kit, 192 Tests
Catalog # CAYM701220-192
Supplier: CAYMAN
Jump to:
Product Details & Documents
For the determination of total calcium concentration in a variety of biological samples, as well as tissue homogenates and cell lysates.
- Measure calcium in plasma, serum, urine, tissue homogenates, and cell lysates
- Assay 40 samples in duplicate
- Measure calcium down to 0,25 mg/dl
- Plate-based colorimetric measurement (570 to 590 nm)
Specifications
Specifications
- Pk:192 Tests
- Description:Calcium Assay Kit
- Cat. No.:CAYM701220-192
- Detection limit:0.25 mg/dl