Order Entry
Slovakia
ContactUsLinkComponent
Glycolic acid ≥99.5% (by acidimetry), AnalaR NORMAPUR® analytical reagent
Glycolic acid ≥99.5% (by acidimetry), AnalaR NORMAPUR® analytical reagent
Catalog # 87061.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Glycolic acid ≥99.5% (by acidimetry), AnalaR NORMAPUR® analytical reagent
Catalog # 87061.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    White to off-white crystals
  • Assay (acidimetric):
    min. 99.5 %
  • Water:
    max. 0.5 %
  • Melting range (lower value):
    min. 75 °C
  • Melting range (upper value):
    max. 79 °C
  • Iron (Fe):
    max. 5 ppm
  • Heavy metals (as Pb):
    max. 10 ppm
  • Identity (IR):
    Passes test

Specification Test Results

About this item

 :
 :76.05 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
 :GHS07
CAS Number:79-14-1
Formula:C₂H₄O₃
MDL Number:MFCD00004312