Pharmaceutical secondary standard, Quercetin, certified reference material, Supelco®
Catalog # PHR1488-1G
Supplier: Merck
CAS Number:
Specifications
- Concentration:Neat
- Pk:1 g
- Standards:Pharmaceutical secondary standard (CRM)
- CAS No.:6151-25-3
- Description:Quercetin
- Description:Quercetin
Specifications
About this item
Traceable to Ph. Eur. Y0001009 and to USP 1592409.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.