Order Entry
Slovakia
ContactUsLinkComponent
Pharmaceutical secondary standard, Phenytoin, certified reference material, Supelco®
Pharmaceutical secondary standard, Phenytoin, certified reference material, Supelco®
Catalog # PHR1139-1G
Supplier:  Merck
CAS Number:  
undefined
Pharmaceutical secondary standard, Phenytoin, certified reference material, Supelco®
Catalog # PHR1139-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1 g
  • Standards:
    Pharmaceutical secondary standard (CRM)
  • CAS No.:
    57-41-0
  • Description:
    Phenytoin
  • Description:
    Phenytoin

Specifications

About this item

Traceable to PhEur P1290000 and to USP 1535008.

Reduces incidence of grand mal seizures; appears to stabilise excitable membranes perhaps through effects on Na⁺, K⁺, and Ca²⁺ channels.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.