Citalopram-D6 hydrobromide solution, certified reference material, Supelco®
C-090-1ML
: Merck
:
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- Description:Citalopram-D6 hydrobromide solution
- Environmentally Preferable:
- Description:Citalopram-D6 hydrobromide solution
- Pk Info:Ampoule of 1 ml
Citalopram is an antidepressant sold under the trade names Celexa® and Cipramil for the treatment of major depression.
Citalopram is a selective serotonin reuptake inhibitor, a class of drugs that also includes fluoxetine, paroxetine, and sertraline. This internal standard is suitable for quantitation of citalopram levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
: Snap-N-Spike®/Snap-N-Shoot®