0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Sulfadimethoxine sodium salt, analytical standard, Supelco®
Sulfadimethoxine sodium salt, analytical standard, Supelco®
S7385-10G
:
Merck
:
Sulfadimethoxine sodium salt, analytical standard, Supelco®
S7385-10G
:
Merck
:
Pk:
10 g
Standards:
analytical standard
CAS No.:
1037-50-9
Description:
Sulfadimethoxine sodium salt
Description:
Sulfadimethoxine sodium salt