Order Entry
Slovakia
ContactUsLinkComponent
Sulfamethizole, >=99% (HPLC), analytical standard, Supelco®
Sulfamethizole, >=99% (HPLC), analytical standard, Supelco®
Catalog # S5632-25G
Supplier:  Merck
CAS Number:  
undefined
Sulfamethizole, >=99% (HPLC), analytical standard, Supelco®
Catalog # S5632-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 g
  • Standards:
    analytical standard
  • CAS No.:
    144-82-1
  • Description:
    Sulfamethizole, ≥99% (HPLC)
  • Description:
    Sulfamethizole

Specifications

About this item